MilliporeSigma
A large solvent isotope effect on protein association thermodynamics.
Biochemistry (2013-08-30)
Christopher Eginton, Dorothy Beckett
ABSTRACT
Solvent reorganization can contribute significantly to the energetics of protein-protein interactions. However, our knowledge of the magnitude of the energetic contribution is limited, in part, by a dearth of quantitative experimental measurements. The biotin repressor forms a homodimer as a prerequisite to DNA binding to repress transcription initiation. At 20 °C, the dimerization reaction, which is thermodynamically coupled to binding of a small ligand, bio-5'-AMP, is characterized by a Gibbs free energy of -7 kcal/mol. This modest net dimerization free energy reflects underlying, very large opposing enthalpic and entropic driving forces of 41 ± 3 and -48 ± 3 kcal/mol, respectively. The thermodynamics have been interpreted as indicating coupling of solvent release to dimerization. In this work, this interpretation has been investigated by measuring the effect of replacing H2O with D2O on the dimerization thermodynamics. Sedimentation equilibrium measurements performed at 20 °C reveal a solvent isotope effect of -1.5 kcal/mol on the Gibbs free energy of dimerization. Analysis of the temperature dependence of the reaction in D2O indicates enthalpic and entropic contributions of 28 and -37 kcal/mol, respectively, considerably smaller than the values measured in H2O. These large solvent isotope perturbations to the thermodynamics are consistent with a significant contribution of solvent release to the dimerization reaction.
Product Description
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Adenosine 2′(3′)-monophosphate mixed isomers
Biotin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material