Merck
  • Perioperative fluid therapy with tetrastarch and gelatin in cardiac surgery--a prospective sequential analysis*.
Perioperative fluid therapy with tetrastarch and gelatin in cardiac surgery--a prospective sequential analysis*.
Critical care medicine (2013-08-28)
Ole Bayer, Daniel Schwarzkopf, Torsten Doenst, Deborah Cook, Bjoern Kabisch, Christoph Schelenz, Michael Bauer, Niels C Riedemann, Yasser Sakr, Matthias Kohl, Konrad Reinhart, Christiane S Hartog
ABSTRACT
To determine clinical outcomes of synthetic colloids, tetrastarch, and gelatin, used before and after cardiac surgery. Prospective observational cohort study. Fifty-bed cardiac ICU. Six thousand four hundred seventy-eight consecutive patients with cardiopulmonary bypass surgery. Fluid therapy in the operating room and on the ICU directed at preset hemodynamic goals: 1) hydroxyethyl starch (predominantly 6% hydroxyethyl starch 130/0.4) in 2004-2006, n = 2,137; 2) 4% gelatin in 2006-2008, n = 2,324; and 3) only crystalloids in 2008-2010, n = 2,017. Renal replacement therapy was more common during periods when patients received synthetic colloids compared to only crystalloids. Risk of renal replacement therapy was greater after hydroxyethyl starch (odds ratio, 2.29; 95% CI, 1.47-3.60) and gelatin (odds ratio, 2.75; 95% CI, 1.84-4.16; both p < 0.001) compared to crystalloid. Propensity score stratification confirmed greater use of renal replacement therapy in the hydroxyethyl starch and gelatin periods compared to the crystalloid period (odds ratio, 1.46 [1.08, 1.97]; p = 0.013 and odds ratio, 1.72 [1.33, 2.24]; p < 0.001, respectively). Time to vasopressor cessation, normalization of serum lactate, and mean arterial pressure did not differ among groups. Total fluid requirement was 163 mL/kg in the hydroxyethyl starch period, 207 mL/kg in the gelatin period, and 224 mL/kg in the crystalloid period. Fluid intake was higher in the crystalloid group only during the first 20 hours. In cardiac surgery patients, fluid therapy with perioperative administration of synthetic colloids carries a high risk of renal replacement therapy and is not more effective than treating with only crystalloids.
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture