MilliporeSigma
  • Malachite green mediates homodimerization of antibody VL domains to form a fluorescent ternary complex with singular symmetric interfaces.
Malachite green mediates homodimerization of antibody VL domains to form a fluorescent ternary complex with singular symmetric interfaces.
Journal of molecular biology (2013-08-28)
Chris Szent-Gyorgyi, Robyn L Stanfield, Susan Andreko, Alison Dempsey, Mushtaq Ahmed, Sarah Capek, Alan S Waggoner, Ian A Wilson, Marcel P Bruchez
ABSTRACT
We report that a symmetric small-molecule ligand mediates the assembly of antibody light chain variable domains (VLs) into a correspondent symmetric ternary complex with novel interfaces. The L5* fluorogen activating protein is a VL domain that binds malachite green (MG) dye to activate intense fluorescence. Crystallography of liganded L5* reveals a 2:1 protein:ligand complex with inclusive C2 symmetry, where MG is almost entirely encapsulated between an antiparallel arrangement of the two VL domains. Unliganded L5* VL domains crystallize as a similar antiparallel VL/VL homodimer. The complementarity-determining regions are spatially oriented to form novel VL/VL and VL/ligand interfaces that tightly constrain a propeller conformer of MG. Binding equilibrium analysis suggests highly cooperative assembly to form a very stable VL/MG/VL complex, such that MG behaves as a strong chemical inducer of dimerization. Fusion of two VL domains into a single protein tightens MG binding over 1000-fold to low picomolar affinity without altering the large binding enthalpy, suggesting that bonding interactions with ligand and restriction of domain movements make independent contributions to binding. Fluorescence activation of a symmetrical fluorogen provides a selection mechanism for the isolation and directed evolution of ternary complexes where unnatural symmetric binding interfaces are favored over canonical antibody interfaces. As exemplified by L5*, these self-reporting complexes may be useful as modulators of protein association or as high-affinity protein tags and capture reagents.
Sigma-Aldrich
Malachite Green solution, for microscopy
Supelco
Malachite Green chloride, analytical standard
Sigma-Aldrich
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Malachite Green oxalate salt, certified by the BSC
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, for microscopy
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade