MilliporeSigma
  • Detection of biothiols in human serum by QDs based flow injection "turn off-on" chemiluminescence analysis system.
Detection of biothiols in human serum by QDs based flow injection "turn off-on" chemiluminescence analysis system.
Talanta (2013-08-21)
Linlin Liu, Qiang Ma, Yang Li, Ziping Liu, Xingguang Su
ABSTRACT
In the present work, a flow injection (FI) "turn off-on" chemiluminescent method was developed for the determination of glutathione (GSH). Strong chemiluminescence (CL) signals were observed from the hydrogen peroxide and CdTe quantum dots (QDs) system under basic condition, addition of trace amount of Cu(Ⅱ) could caused significant CL quenching of the CdTe QDs-H2O2 system. In the presence of biothiols, Cu(Ⅱ) can be removed from CdTe QDs surface via forming Cu(Ⅱ)-S bond with thiols, and the CL signal of CdTe QDs-H2O2 system was recovered. Thus, the CL signals of CdTe QDs-H2O2 system were turned off and on by the addition of Cu(Ⅱ) and biothiols respectively, and a flow injection CL analysis system for the determination of biothiols was established. Under the optimum conditions, the CL intensity and the concentration of GSH have a good linear relationship in the range of 2.0×10(-9)-6.5×10(-7) mol L(-1) (R(2)=0.9993). The limit of detection for GSH is 1.5×10(-9) mol L(-1) (S/N=3). This method has been applied to detect GSH in human serum with satisfactory results.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Tellurium, shot, ≤2 mm, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, granular, −5-+50 mesh, 99.99% trace metals basis
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Tellurium, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
Tellurium, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Tellurium, pieces, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 8-20 mesh