MilliporeSigma
  • Global DNA hypermethylation is associated with high serum levels of persistent organic pollutants in an elderly population.
Global DNA hypermethylation is associated with high serum levels of persistent organic pollutants in an elderly population.
Environment international (2013-08-13)
Lars Lind, Johanna Penell, Karin Luttropp, Louise Nordfors, Anne-Christine Syvänen, Tomas Axelsson, Samira Salihovic, Bert van Bavel, Tove Fall, Erik Ingelsson, P Monica Lind
ABSTRACT
Dioxin exposure has experimentally been associated with changes in DNA methylation, an epigenetic change that is associated with disease. The present study aims to investigate if serum levels of dioxin and other persistent environmental pollutants are related to global DNA methylation in a human sample. In the population-based Prospective Investigation of the Vasculature in Uppsala Seniors (PIVUS) study (all aged 70), global DNA methylation was measured by the Luminometric Methylation Assay in 524 subjects. Twenty-three different POPs, including 16 PCBs, five pesticides, one dioxin (OCDD) and one brominated flame retardant (BDE47) were analysed by HRGC/HRMS. Ten single nucleotide polymorphisms (SNPs) in the Aryl hydrocarbon (Ah)-receptor were analysed by mini-sequencing. High levels of toxic equivalency (TEQ) for PCBs and dioxin were associated with DNA hypermethylation (p=0.030). This was mainly attributed to coplanar non-ortho PCBs. While no significant associations were found between DNA methylation and SNPs in the Ah-receptor, an interaction was found between the SNP rs2237297 and TEQ so that TEQ was associated with hypermethylation (p=0.009) only in subjects with one G-allele (n=103). Also high levels of the PCB126 congener, the OCDD, and the pesticide metabolite p,p'-DDE were related to DNA hypermethylation (p=0.01, 0.03 and 0.003, respectively). In conclusion, in a sample of elderly subjects, high TEQ including PCBs and the dioxin OCDD and high serum levels of PCB126, OCDD, and p,p'-DDE were related to global DNA hypermethylation in a cross-sectional analysis.
Product Description
Supelco
4,4′-DDE, PESTANAL®, analytical standard