MilliporeSigma
  • Dissection of activation parameters in the bell-shaped α-effect following solvent modulation (DMSO-H2O media).
Dissection of activation parameters in the bell-shaped α-effect following solvent modulation (DMSO-H2O media).
The Journal of organic chemistry (2013-08-07)
Ik-Hwan Um, Ji-Sun Kang, Min-Young Kim, Erwin Buncel
ABSTRACT
This paper comprises results of our investigation of the α-effect phenomenon for the reaction of O-p-nitrophenyl thionobenzoate (PNPTB) with butane-2,3-dione monoximate (Ox(-), α-nucleophile) and p-chlorophenoxide (p-ClPhO(-), normal-nucleophile) in DMSO-H2O mixtures of varying compositions at 15.0 °C, 25.0 °C, and 35.0 °C. The reactivity of Ox(-) and p-ClPhO(-) increases significantly as the DMSO content in the medium increases, although the effects of medium on reactivity are not the same for the reactions with Ox(-) and p-ClPhO(-). Ox(-) exhibits the α-effect in all solvent compositions and temperatures. The α-effect increases up to 50 mol % DMSO and then decreases thereafter, resulting in a bell-shaped α-effect profile. Dissection of the activation parameters (i.e., ΔH(‡) and TΔS(‡)) has revealed that the bell-shaped α-effect behavior is due to entropy of activation differences rather than enthalpy terms, although the enthalpy term controls almost entirely the solvent dependence of the reaction rate. Differences in the transition-state (TS) structures for the reactions with Ox(-) (a six-membered cyclic TS) and p-ClPhO(-) (an acyclic TS) are consistent with the entropy-dependent α-effect behavior.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
4-Chlorophenol, ≥99%
Sigma-Aldrich
2,3-Butanedione, 97%
Supelco
2,3-Butanedione, analytical standard
Supelco
Dimethyl Sulfoxide, Pharmaceutical Secondary Standard; Certified Reference Material