MilliporeSigma
  • Functionalization of linen/cotton pigment prints using inorganic nano structure materials.
Functionalization of linen/cotton pigment prints using inorganic nano structure materials.
Carbohydrate polymers (2013-08-06)
N A Ibahim, B M Eid, E Abd El-Aziz, T M Abou Elmaaty
ABSTRACT
The present work opens up a novel strategy for the development of new multifunctional cellulosic pigment prints. The developed process aims at modifying the solvent-free pigment printing formulations via inclusion of certain inorganic nano materials namely silver (Ag-NPs), zinc oxide (ZnO-NPs), zirconium oxide (ZrO₂-NPs) or titanium dioxide (TiO₂-NPs) at 20 g/kg paste followed by screen printing and microwave fixation. The imparted functional properties together with the depth of the obtained prints are governed by the type of nano additives, type of binder and the pigment colorant. The imparted antibacterial and/or UV protection properties to the pigment prints were retained with an acceptable level (>70%) of durability even after 20 washing cycles. The presence of nano materials on the surface of the obtained pigment prints was confirmed using SEM images and EDX spectra.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis