MilliporeSigma
  • A novel biodegradable β-cyclodextrin-based hydrogel for the removal of heavy metal ions.
A novel biodegradable β-cyclodextrin-based hydrogel for the removal of heavy metal ions.
Carbohydrate polymers (2013-08-06)
Zhanhua Huang, Qinglin Wu, Shouxin Liu, Tian Liu, Bin Zhang
ABSTRACT
A novel biodegradable β-cyclodextrin-based gel (CAM) was prepared and applied to the removal of Cd(2+), Pb(2+) and Cu(2+) ions from aqueous solutions. CAM hydrogel has a typical three-dimensional network structure, and showed excellent capability for the removal of heavy metal ions. The effect of different experimental parameters, such as initial pH, adsorbent dosage and initial metal ion concentration, were investigated. The adsorption isotherm data fitted well to the Freundlich model. The adsorption capacity was in the order Pb(2+)>Cu(2+)>Cd(2+) under the same experimental conditions. The maximum adsorption capacities for the metal ions in terms of mg/g of dry gel were 210.6 for Pb(2+), 116.41 for Cu(2+), and 98.88 for Cd(2+). The biodegradation efficiency of the resin reached 79.4% for Gloeophyllum trabeum. The high adsorption capacity and kinetics results indicate that CAM can be used as an alternative adsorbent to remove heavy metals from aqueous solution.
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor