MilliporeSigma
  • Drug delivery systems using sandwich configurations of electrospun poly(lactic acid) nanofiber membranes and ibuprofen.
Drug delivery systems using sandwich configurations of electrospun poly(lactic acid) nanofiber membranes and ibuprofen.
Materials science & engineering. C, Materials for biological applications (2013-08-06)
Ana Paula Serafini Immich, Manuel Lis Arias, Núria Carreras, Rafael Luís Boemo, José Antonio Tornero
ABSTRACT
The primary advantages of electrospun membranes include the ability to obtain very thin fibers that are on the order of magnitude of several nanometers with a considerable superficial area and the possibility for these membranes to be manipulated and processed for many different applications. The purpose of this study is to evaluate and quantify the transport mechanisms that control the release of drugs from polymer-based sandwich membranes produced using the electrospinning processes. These electrospun membranes were composed of poly(lactic acid) (PLA) because it is one of the most promising biodegradable polymers due to its mechanical properties, thermoplastic processability and biological properties, such as its biocompatibility and biodegradability. The transport mechanism that controls the drug delivery was evaluated via the release kinetics of a bioactive agent in physiological serum, which was used as a corporal fluid simulation. To describe the delivery process, mathematical models, such as the Power Law, the classical Higuchi equation and an approach to Fick's Second Law were used. Using the applied mathematical models, it is possible to conclude that control over the release of the drug is significantly dependent on the thickness of the membrane rather than the concentration of the drug.
Product Description
Supelco
Ibuprofen
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Sigma-Aldrich
Polylactic acid, Mw ~60,000