MilliporeSigma
  • Daily acyclovir to decrease herpes simplex virus type 2 (HSV-2) transmission from HSV-2/HIV-1 coinfected persons: a randomized controlled trial.
Daily acyclovir to decrease herpes simplex virus type 2 (HSV-2) transmission from HSV-2/HIV-1 coinfected persons: a randomized controlled trial.
The Journal of infectious diseases (2013-08-01)
Andrew Mujugira, Amalia S Magaret, Connie Celum, Jared M Baeten, Jairam R Lingappa, Rhoda Ashley Morrow, Kenneth H Fife, Sinead Delany-Moretlwe, Guy de Bruyn, Elizabeth A Bukusi, Etienne Karita, Saidi Kapiga, Lawrence Corey, Anna Wald
ABSTRACT
Daily suppressive therapy with valacyclovir reduces risk of sexual transmission of herpes simplex virus type 2 (HSV-2) in HSV-2-serodiscordant heterosexual couples by 48%. Whether suppressive therapy reduces HSV-2 transmission from persons coinfected with HSV-2 and human immunodeficiency virus type 1 (HIV-1) is unknown. Within a randomized trial of daily acyclovir 400 mg twice daily in African HIV-1 serodiscordant couples, in which the HIV-1-infected partner was HSV-2 seropositive, we identified partnerships in which HIV-1-susceptible partners were HSV-2 seronegative to estimate the effect of acyclovir on risk of HSV-2 transmission. We randomly assigned 911 HSV-2/HIV-1-serodiscordant couples to daily receipt of acyclovir or placebo. We observed 68 HSV-2 seroconversions, 40 and 28 in acyclovir and placebo groups, respectively (HSV-2 incidence, 5.1 cases per 100 person-years; hazard ratio [HR], 1.35 [95% confidence interval, .83-2.20]; P = .22). Among HSV-2-susceptible women, vaginal drying practices (adjusted HR, 44.35; P = .004) and unprotected sex (adjusted HR, 9.91; P = .002) were significant risk factors for HSV-2 acquisition; having more children was protective (adjusted HR, 0.47 per additional child; P = .012). Among HSV-2-susceptible men, only age ≤30 years was associated with increased risk of HSV-2 acquisition (P = .016). Treatment of African HSV-2/HIV-1-infected persons with daily suppressive acyclovir did not decrease risk of HSV-2 transmission to susceptible partners. More-effective prevention strategies to reduce HSV-2 transmission from HIV-1-infected persons are needed.
Product Description
Aciclovir, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material