MilliporeSigma
  • Enhancement of protective immune response to recombinant Toxoplasma gondii ROP18 antigen by ginsenoside Re.
Enhancement of protective immune response to recombinant Toxoplasma gondii ROP18 antigen by ginsenoside Re.
Experimental parasitology (2013-07-31)
Daofeng Qu, Jianzhong Han, Aifang Du
ABSTRACT
Toxoplasma gondii, the etiological agent of toxoplasmosis, is an obligate intracellular protozoan parasite that infects a variety of mammals including humans. In an attempt to find new antigen-adjuvant combinations that enhance the immunogenicity of antigen candidates for toxoplasma vaccines, we analyzed the potent protection in mice immunized with recombinant protein ROP18 when co-administered with ginsenoside Re, a most important component isolated from Panax ginseng. All immunized mice produced specific anti-rROP18 immunoglobulins, with high levels of IgG antibody and a mixed IgG1/IgG2a response, with predominance of IgG1 production. The cellular and humoral immune responses were associated with the production of IFN-γ and IL-4 cytokines respectively. Vaccinated mice displayed a significantly increased survival time compared with control mice which died within 6 days of challenge with RH strain. Our data demonstrate that by addition of ginsenoside Re, the rROP18 triggered a stronger humoral and cellular response against T. gondii, and that Re is a promising vaccine adjuvant against toxoplasmosis, deserves further evaluation and development.