MilliporeSigma
  • Feasibility of using low density polyethylene sheets to detect atmospheric organochlorine pesticides in Alexandria, Egypt.
Feasibility of using low density polyethylene sheets to detect atmospheric organochlorine pesticides in Alexandria, Egypt.
Environmental pollution (Barking, Essex : 1987) (2013-07-23)
Mohammed A Khairy, Rainer Lohmann
ABSTRACT
Egypt is a major agricultural country in Africa with a known past of organochlorine pesticides (OCPs) application, yet data on atmospheric levels of OCPs in Egypt is sparse. Low density polyethylene (LDPE) passive samplers were therefore deployed for 3 weeks each at 11 locations in July, 2010 and January, 2011 in Alexandria to screen for gas-phase OCPs. Performance reference compounds were used to investigate the uptake kinetics. Field-derived sampler-air partitioning coefficients (KPE-As) for OCPs were significantly correlated against the compounds' subcooled liquid vapor pressure (log PL): [log KPE-A = -0.77 ± 0.07 log PL + 6.35 ± 0.13 (R(2) = 0.90; n = 17; SE = 0.19; p < 0.001)]. Estimated and measured OCP concentrations in Alexandria agreed well (factor difference ≤ 2) indicating the feasibility of monitoring OCPs using LDPEs. OCP concentrations ranged from <LOD to 168 pg/m(3). Calculated isomeric ratios indicated recent usage of chlordanes and endosulfans.
Product Description
Polyethylene (LDPE), ERM®, certified reference material
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC