MilliporeSigma
  • One-pot, water-based and high-yield synthesis of tetrahedral palladium nanocrystal decorated graphene.
One-pot, water-based and high-yield synthesis of tetrahedral palladium nanocrystal decorated graphene.
Nanoscale (2013-07-19)
Gengtao Fu, Lin Tao, Min Zhang, Yu Chen, Yawen Tang, Jun Lin, Tianhong Lu
ABSTRACT
This paper reports a facile, water-based and one-pot synthesis of tetrahedral Pd nanocrystals (Pd-TNPs) with high yield and good size monodispersity supported on reduced graphene oxide (RGO) nanosheets via a co-chemical reduction method. The key synthetic strategy employed a positively charged polyallylamine-Pd(II) complex (PAH-Pd(II)) with un-coordinated amine groups as a linker molecule to immobilize Pd(II) species on the negatively charged graphene oxide (GO) surface through electrostatic interaction. As characterized by transmission electron microscopy (TEM), energy dispersive spectroscopy (EDS), X-ray diffraction (XRD), Raman spectroscopy, thermogravimetric analysis (TGA), X-ray photoelectron spectroscopy (XPS) techniques, well-defined Pd-TNPs with an average size of 9 nm were uniformly distributed on the RGO surface. The as-prepared Pd-TNPs/RGO nanohybrid with excellent colloidal stability in aqueous solution exhibits superior catalytic activity towards the degradation of methylene blue (MB) compared to both unsupported Pd-TNPs and Pd black. Thus, the resultant Pd-TNPs/RGO nanohybrid, as a promising heterogeneous catalyst, might have wide potential applications in water-based catalysis systems for the future.
Sigma-Aldrich
Poly(allylamine) solution, average Mw ~65,000, 10 wt. % in H2O
Sigma-Aldrich
Poly(allylamine hydrochloride), average Mw 50,000
Sigma-Aldrich
Poly(allylamine hydrochloride), average Mw ~17,500 (GPC vs. PEG std.)
Sigma-Aldrich
Poly(allylamine) solution, 20 wt. % in H2O