MilliporeSigma
  • Antifungal property of hibicuslide C and its membrane-active mechanism in Candida albicans.
Antifungal property of hibicuslide C and its membrane-active mechanism in Candida albicans.
Biochimie (2013-07-03)
Ji Hong Hwang, Qinglong Jin, Eun-Rhan Woo, Dong Gun Lee
ABSTRACT
In this study, the antifungal activity and mode of action(s) of hibicuslide C derived from Abutilon theophrasti were investigated. Antifungal susceptibility testing showed that hibicuslide C possessed potent activities toward various fungal strains and less hemolytic activity than amphotericin B. To understand the antifungal mechanism(s) of hibicuslide C in Candida albicans, flow cytometric analysis with propidium iodide was done. The results showed that hibicuslide C perturbed the plasma membrane of the C. albicans. The analysis of the transmembrane electrical potential with 3,3'-dipropylthiacarbocyanine iodide [DiSC3(5)] indicated that hibicuslide C induced membrane depolarization. Furthermore, model membrane studies were performed with calcein encapsulating large unilamellar vesicles (LUVs) and FITC-dextran (FD) loaded LUVs. These results demonstrated that the antifungal effects of hibicuslide C on the fungal plasma membrane were through the formation of pores with radii between 2.3 nm and 3.3 nm. Finally, in three dimensional flow cytometric contour plots, a reduced cell sizes by the pore-forming action of hibicuslide C were observed. Therefore, the present study suggests that hibicuslide C exerts its antifungal effect by membrane-active mechanism.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Dextran, analytical standard, for GPC, 150,000
Supelco
Dextran, analytical standard, for GPC, 1,000
Supelco
Dextran, analytical standard, for GPC, 5,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Supelco
Dextran, analytical standard, for GPC, 80,000
Supelco
Dextran, analytical standard, for GPC, 25,000
Supelco
Dextran, analytical standard, for GPC, 12,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000