MilliporeSigma
  • New rapid scheme for distinguishing the subspecies of the Mycobacterium abscessus group and identifying Mycobacterium massiliense isolates with inducible clarithromycin resistance.
New rapid scheme for distinguishing the subspecies of the Mycobacterium abscessus group and identifying Mycobacterium massiliense isolates with inducible clarithromycin resistance.
Journal of clinical microbiology (2013-06-28)
Shamira J Shallom, Paul J Gardina, Timothy G Myers, Yinong Sebastian, Patricia Conville, Leslie B Calhoun, Hervé Tettelin, Kenneth N Olivier, Gulbu Uzel, Elizabeth P Sampaio, Steven M Holland, Adrian M Zelazny
ABSTRACT
Mycobacterium abscessus (M. abscessus sensu lato, or the M. abscessus group) comprises three closely related taxa whose taxonomic statuses are under revision, i.e., M. abscessus sensu stricto, Mycobacterium bolletii, and Mycobacterium massiliense. We describe here a simple, robust, and cost-effective PCR-based method for distinguishing among M. abscessus, M. massiliense, and M. bolletii. Based on the M. abscessus ATCC 19977(T) genome, regions that discriminated between M. abscessus and M. massiliense were identified through array-based comparative genomic hybridization. A typing scheme using PCR primers designed for four of these locations was applied to 46 well-characterized clinical isolates comprising 29 M. abscessus, 15 M. massiliense, and 2 M. bolletii isolates previously identified by multitarget sequencing. Interestingly, 2 isolates unequivocally identified as M. massiliense were shown to have a full-length erm(41) gene instead of the expected gene deletion and showed inducible clarithromycin resistance after 14 days. We propose using this PCR-based typing scheme combined with erm(41) PCR for straightforward identification of M. abscessus, M. massiliense, and M. bolletii and the assessment of inducible clarithromycin resistance. This method can be easily integrated into a routine workflow to provide subspecies-level identification within 24 h after isolation of the M. abscessus group.
Product Description
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Pricing and availability is not currently available.