MilliporeSigma
  • Monolithic poly(N-vinylcarbazole-co-1,4-divinylbenzene) capillary columns for the separation of biomolecules.
Monolithic poly(N-vinylcarbazole-co-1,4-divinylbenzene) capillary columns for the separation of biomolecules.
The Analyst (2013-06-27)
Rainer Koeck, Rania Bakry, Richard Tessadri, Guenther K Bonn
ABSTRACT
Monolithic capillary columns were prepared by thermally initiated free radical copolymerization of N-vinylcarbazole (NVC) and 1,4-divinylbenzene (DVB) within the confines of 200 and 100 μm i.d. fused silica capillaries. The reaction was carried out under the influence of inert micro-(toluene) and macroporogen (1-decanol) and α,α'-azoisobutyronitrile (AIBN) as a free radical initiator. The material proved high mechanical stability applying water and acetonitrile as mobile phases. The morphological and porous properties were studied by scanning electron microscopy (SEM), nitrogen sorption (BET) and mercury intrusion porosimetry (MIP). The homogeneity of the copolymerization process was confirmed by elemental analysis and monomer conversion measurements. The newly developed NVC/DVB monolithic supports showed high separation efficiency towards biomolecules, applying reversed-phase (RP) and ion-pair reversed-phase (IP-RP) separation modes, which is exemplified by the separations of peptides, proteins and oligonucleotides. Furthermore the maximum loading capacity was evaluated. The chromatographic performance under isocratic elution was determined in terms of theoretical plate number and plate height, where up to 41,000 plates per column and a minimum plate height value of 1.7 μm were achieved, applying oligonucleotide separations. In gradient elution mode, peak capacities of 96 and 127 were achieved within a gradient time window of 60 min for protein and oligonucleotide separations, respectively. The material proved to have high permeability, good repeatability of the fabrication process and high surface areas in the range of 120-160 m(2) g(-1).
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mn 25,000-50,000
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mw ~1,100,000, powder
Sigma-Aldrich
Divinylbenzene, technical grade, 80%