MilliporeSigma
  • The effect of rocuronium and sugammadex on neuromuscular blockade in a child with congenital myotonic dystrophy type 1.
The effect of rocuronium and sugammadex on neuromuscular blockade in a child with congenital myotonic dystrophy type 1.
Paediatric anaesthesia (2013-06-15)
Amelia Pickard, Clinton Lobo, Peter A Stoddart
ABSTRACT
Myotonic dystrophy type 1 (MD1) is the commonest muscular dystrophy found in adults; however, it may present in the neonatal period with hypotonia, talipes, poor feeding, and respiratory failure. Inheritance is autosomal dominant with a defect in the DMPK gene found on the long arm of chromosome 19 with variable expansion of the cytosine-thymine-guanine (CTG) triplet repeat. A 14-month-old boy with congenital MD type 1 was scheduled for percutaneous endoscopic gastrostomy (PEG) insertion, orchidopexy, and division of tongue-tie. Following induction of anesthesia, acceleromyography was used to monitor neuromuscular function. This revealed a very rapid onset of profound neuromuscular block which lasted significantly longer than would be expected in a child without MD1. Sugammadex reversed the block rapidly. The anesthetic management of children with MD1 has been well described but not the acceleromyographic monitored use of rocuronium and its subsequent reversal with the new cyclodextrin sugammadex.
Product Description
Rocuronium for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rocuronium bromide, ≥97% (perchloric acid titration)