MilliporeSigma
  • In vivo analgesic and anti-inflammatory activities of Rosmarinus officinalis aqueous extracts, rosmarinic acid and its acetyl ester derivative.
In vivo analgesic and anti-inflammatory activities of Rosmarinus officinalis aqueous extracts, rosmarinic acid and its acetyl ester derivative.
Pharmaceutical biology (2013-06-07)
Rodrigo Lucarini, Wagner A Bernardes, Daniele S Ferreira, Marcos G Tozatti, Ricardo Furtado, Jairo K Bastos, Patrícia M Pauletti, Ana H Januário, Márcio L Andrade E Silva, Wilson R Cunha
ABSTRACT
Despite several pharmacological applications of Rosmarinus officinalis L. (Lamiaceae), studies on its analgesic and anti-inflammatory properties have been scarce. The aim of this work was to use in vivo models to evaluate the analgesic and anti-inflammatory activities of the aqueous extracts obtained from leaves (AEL) and stems (AES) of Rosmarinus officinalis, as well as its isolated compound--rosmarinic acid (RA). We also prepared and assessed the acetyl ester derivative of RA. The analgesic activity was evaluated using abdominal constriction and formalin tests. For the evaluation of the anti-inflammatory effects, carrageenin-induced paw edema in rats were used. The extracts were used at doses of 100, 200 and 400 mg kg⁻¹ compounds were tested at 10, 20 and 40 mg kg⁻¹. Orally administered AEL, AES and RA were not significantly active at any of the doses tested during the abdominal constriction test; the acetyl ester derivative of RA displayed significant analgesic activity. In the carrageenin-induced paw edema assay, the acetyl derivative of RA at all the tested doses produced significant anti-inflammatory effects and reduced the number of paw licks in the second phase of the formalin test. The results suggest that the analgesic effects of the acetyl derivative of RA operate via a peripheral-mediated mechanism. The acetyl ester derivative of RA is potentially applicable as a new lead compound for the management of pain and inflammation.
Sigma-Aldrich
Rosmarinic acid, 96%
Rosmarinic acid, primary reference standard
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.