MilliporeSigma
  • Effects of particle size, food, and capsule shell composition on the oral bioavailability of dabrafenib, a BRAF inhibitor, in patients with BRAF mutation-positive tumors.
Effects of particle size, food, and capsule shell composition on the oral bioavailability of dabrafenib, a BRAF inhibitor, in patients with BRAF mutation-positive tumors.
Journal of pharmaceutical sciences (2013-04-24)
Daniele Ouellet, Kenneth F Grossmann, Giselle Limentani, Noelia Nebot, Kevin Lan, Lara Knowles, Michael S Gordon, Sunil Sharma, Jeffrey R Infante, Patricia M Lorusso, Girish Pande, Elizabeth C Krachey, Samuel C Blackman, Stanley W Carson
ABSTRACT
Dabrafenib is a small-molecule inhibitor of BRAF kinase activity that is currently being developed for the treatment of BRAF V600 mutation-positive melanoma. This clinical, open-label, two-cohort (n = 14 per cohort), randomized study was designed to evaluate the effect of drug substance particle size, and food on the plasma pharmacokinetics of a single oral dose of dabrafenib in patients with BRAF V600 mutation-positive solid tumors. In addition, an exploratory cross-cohort comparison of the relative bioavailability of single-dose dabrafenib administered in gelatin and hydroxypropyl methylcellulose (HPMC) capsules was performed. Higher bioavailability was noted with nonmicronized drug substance (larger particle size), under fasting conditions, and with HPMC capsules. Initial dissolution results at pH 1.2 showed higher dissolution of gelatin relative to HPMC capsules inconsistent with clinical data. Subsequent in vitro dissolution studies were conducted in fasted-state simulated gastric fluid over a 24-h period and showed that HPMC capsules reached a higher percentage of dabrafenib dissolved than gelatin capsules. The presence of HPMC is believed to inhibit precipitation of dabrafenib as the freebase, thereby maintaining a supersaturated solution over an extended period of time. Dabrafenib has been administered in pivotal clinical studies on an empty stomach using micronized drug substance in HPMC capsules.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (methoxyl group, on Dry Basis), meets USP testing specifications, viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)