MilliporeSigma
  • Hydrophilic versus hydrophobic porogens for engineering of poly(lactide-co-glycolide) microparticles containing risedronate sodium.
Hydrophilic versus hydrophobic porogens for engineering of poly(lactide-co-glycolide) microparticles containing risedronate sodium.
Pharmaceutical development and technology (2012-06-05)
Maha Nasr, Gehanne A S Awad, Samar Mansour, Abdelhamid Al Shamy, Nahed D Mortada
ABSTRACT
The aim of this study was to investigate the effect of two mechanistically different porogens, namely: the hydrophilic hydroxy-propyl-β-cyclodextrin and the hydrophobic porogens (mineral oil and corn oil) in producing open/closed pored engineered polylactide-co-glycolic-acid microspheres suitable for pulmonary delivery of risedronate sodium (RS). Surface morphology of the microspheres was studied and they were characterized for entrapment efficiency (%EE), particle size, and porosity as well as aerodynamic and flow properties. Selected formulae were investigated for in vitro drug release and deposition behavior using next generation impactor. Furthermore, the safety of the free drug and the selected prepared systems was assessed by MTT viability test performed on Calu-3 cell line. The current work revealed that HP-β-CD produced open-pored microspheres, while oils produced closed pored microspheres. Modulation of preparation parameters generated porous RS microspheres with high %EE, sustained drug release profile up to 15 days, suitable geometric and aerodynamic particle sizes and excellent flow properties. The safety of HP-β-CD systems was higher than the systems utilizing oil as porogen. Porogen type affected the behavior of the microspheres as demonstrated by the various characterization experiments, with microspheres prepared using HP-β-CD being superior to those prepared using oils as porogens.
Sigma-Aldrich
β-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
β-Cyclodextrin, ≥97%
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Etidronate disodium hydrate, ≥97% (NMR), solid
Sigma-Aldrich
β-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000
Sigma-Aldrich
β-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% (HPLC)
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Supelco
Corn oil, analytical standard
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000