MilliporeSigma
  • High efficiency method to obtain supercoiled DNA with a commercial plasmid purification kit.
High efficiency method to obtain supercoiled DNA with a commercial plasmid purification kit.
Acta biochimica Polonica (2012-06-14)
Antonietta Carbone, Flavia Marialucia Fioretti, Laura Fucci, Juan Ausió, Marina Piscopo
ABSTRACT
Supercoiled state corresponds to the active form for plasmid applications. The relaxed circular form of plasmids is often inactive or poorly active. To obtain significant amounts of almost fully supercoiled DNA, we modified the standard protocol of a commercially available Qiagen plasmid purification kit. Our changes led to isolation of almost 100% of the plasmids in the supercoiled state. The modified protocol was used to purify different plasmids with consistent results. The purified plasmids maintain supercoiled state for about two months. The modified protocol is very advantageous because it allows easy DNA production with high degree of supercoiled form at low cost.
Sigma-Aldrich
GenElute HP Plasmid Miniprep Kit, sufficient for 70 preparations
Sigma-Aldrich
GenElute HP Plasmid Miniprep Kit, sufficient for 350 preparations