MilliporeSigma
  • DDX39, upregulated in lung squamous cell cancer, displays RNA helicase activities and promotes cancer cell growth.
DDX39, upregulated in lung squamous cell cancer, displays RNA helicase activities and promotes cancer cell growth.
Cancer biology & therapy (2007-06-06)
Takeyuki Sugiura, Yuki Nagano, Yoshie Noguchi
ABSTRACT
To explore differentially expressed genes involved in non-small cell lung cancer progression, we used the gene expression profile database of various human tissues and identified DDX39, a new member of the DEAD box RNA helicases, showing overexpression in human lung squamous cell carcinoma (LSCC) but not in lung adenocarcinoma (LAC). There existed three types of alternatively spliced DDX39 variants (DDX39-L, -S and -SS), of which only DDX39-L contains all the motifs required for RNA helicase activity. RT-PCR analysis verified the increased expression of DDX39-L in LSCC, but not LAC, cultured cells compared with normal bronchial epithelial cells. A high sequence similarity to UAP56 and punctate nuclear localization pattern of DDX39-L suggest that it plays a role in RNA splicing/export. Recombinant DDX39-L binds RNA, hydrolyzes NTPs in an RNA-dependent manner and unwinds double strand RNA bidirectionally, proving that DDX39 is an RNA helicase. Overexpression of DDX39-L stimulates colony formation of HeLa cells, probably through elevation of a translational level, indicating the biological significance of DDX39 in cancer pathogenesis. Thus, DDX39 is a novel RNA helicase capable of promoting cancer cell growth and, thereby, can be a potential target for development of a therapeutic strategy for LSCC.
Sigma-Aldrich
Anti-DDX39 antibody produced in rabbit, IgG fraction of antiserum