MilliporeSigma
  • Unexpected rearrangement of enantiomerically pure 3-aminoquinuclidine as a simple way of preparing diastereomeric octahydropyrrolo[2,3-c]pyridine derivatives.
Unexpected rearrangement of enantiomerically pure 3-aminoquinuclidine as a simple way of preparing diastereomeric octahydropyrrolo[2,3-c]pyridine derivatives.
Chirality (2008-09-17)
Igor Goljer, Albert Molinari, Yanan He, Lisa Nogle, Weilin Sun, Brandon Campbell, Oliver McConnell
ABSTRACT
Reaction of (S)- or (R)-3-aminoquinuclidine with 2-chloropyrimidine or 2-bromopyrimidine led to an unexpected formation of both cis- and trans-octahydropyrrolo [2,3]pyridine derivatives. A single-step synthesis of two of the four stereoisomers of these octahydropyrrolo[2,3]pyridine derivatives provides a convenient way of generating stereochemically defined isomers. Optimization of reaction conditions was carried out by (1)H NMR monitoring. The relative and absolute stereochemistry of all four stereoisomers was determined by a combination of (1)H, (13)C, and (15)N NMR spectroscopy and vibrational circular dichroism spectroscopy.
Sigma-Aldrich
2-Chloropyrimidine, 95%