MilliporeSigma
  • A new fluorescence method for the continuous determination of surface lipid oxidation in lipoproteins and plasma.
A new fluorescence method for the continuous determination of surface lipid oxidation in lipoproteins and plasma.
Free radical research (1995-10-01)
G Hofer, D Lichtenberg, A Hermetter
ABSTRACT
We report on a new method for the determination of lipid oxidation in lipoproteins and plasma. The biological lipid system is preloaded with a fluorescent analog of phosphatidylcholine containing diphenylhexatriene (DPH) propionic acid covalently linked to the sn-2 position. When externally added, the respective phospholipid label (DPHPC) localizes to the surface monolayer of a lipoprotein. Under oxidative conditions (e.g. in the presence of Cu2+ ions) the fluorophore undergoes decomposition, resulting in a continuous decrease of fluorescence intensity which reflects the oxidation of a chemically defined phospholipid molecule with well defined localization. When incorporated into LDL particles, the kinetics of the decrease in DPHPC fluorescence intensity upon exposure to Cu2+ us very similar to that of conjugated diene accumulation. Furthermore, our assay can be applied to follow the oxidation of lipids in diluted serum and may also be developed into a suitable test system for clinical studies of susceptibility of plasma lipids to oxidation.
Sigma-Aldrich
Lipase Substrate
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase Substrate, ≥95% (HPLC)