MilliporeSigma
  • Facile synthesis of alpha,alpha-diisobutylglycine and anchoring its derivatives onto PAL-PEG-PS resin.
Facile synthesis of alpha,alpha-diisobutylglycine and anchoring its derivatives onto PAL-PEG-PS resin.
The Journal of organic chemistry (2003-12-06)
Yanwen Fu, Marcus A Etienne, Robert P Hammer
ABSTRACT
alpha,alpha-Diisobutylglycine has been synthesized using a Pd-mediated dialkylation of ethyl nitroacetate as a key first step. The free alphaalphaAA is N(alpha)-protected and has been applied to the assembly of conformationally constrained peptide analogues. Mixed anhydrides from BOP-Cl and Fmoc-alphaalphaAA-OH are used for anchoring alphaalphaAAs onto a trialkoxybenzyl linker on PEG-PS grafted support, upon which a beta-strand mimic with difficult sequence is assembled in a superior quality.
Sigma-Aldrich
Ethyl nitroacetate, 97%