MilliporeSigma
  • 1, 2-Diolates of titanium as suitable precursors for the preparation of photoactive high surface titania.
1, 2-Diolates of titanium as suitable precursors for the preparation of photoactive high surface titania.
Applied Catalysis. B, Environmental (1999)
Saadoun L, et al.