MilliporeSigma
Salubrinal reduces expression and activity of MMP13 in chondrocytes.
Osteoarthritis and cartilage (2013-03-12)
K Hamamura, C-C Lin, H Yokota
ABSTRACT
Stress to the endoplasmic reticulum (ER) and inflammatory cytokines induce expression and activity of matrix metalloproteinase 13 (MMP13). Since a synthetic agent, salubrinal, is known to alleviate ER stress and attenuate nuclear factor kappa B (NFκB) signaling, we addressed a question whether upregulation of MMP13 by ER stress and cytokines is suppressed by administration of salubrinal. Using C28/I2 human chondrocytes, we applied ER stress with tunicamycin and inflammatory distress with tumor necrosis factor α (TNFα) and interleukin 1β (IL1β). RNA interference with siRNA specific to NFκB p65 (RelA) was employed to examine a potential involvement of NFκB signaling in salubrinal's action in regulation of MMP13. We also employed primary human chondrocytes and evaluated MMP13 activity. The result showed that tunicamycin activated p38 mitogen-activated protein kinase (MAPK), while inflammatory cytokines activated p38 MAPK and NFκB. In both cases, salubrinal significantly reduced expression and activity of MMP13. Silencing NFκB reduced inflammatory cytokine-driven upregulation of MMP13 activity. The results demonstrate that salubrinal downregulates expression and activity MMP13 through p38 and NFκB signaling, suggesting its potential usage to treat degenerative diseases such as osteoarthritis.
Sigma-Aldrich
Salubrinal, ≥98% (HPLC)