- Notable coordination effects of 2-pyridinesulfonamides leading to efficient aziridination and selective aziridine ring opening.
Notable coordination effects of 2-pyridinesulfonamides leading to efficient aziridination and selective aziridine ring opening.
Organic letters (2004-10-22)
Hoon Han, Imhyuck Bae, Eun Jeong Yoo, Junseong Lee, Youngkyu Do, Sukbok Chang
PMID15496111
ABSTRACT
[reaction: see text] We have developed, on the basis of a chelation-strategy, an efficient copper-catalyzed aziridination protocol with the use of 5-methyl-2-pyridinesulfonamide and PhI(OAc)(2). The reaction proceeds smoothly under mild conditions to give aziridines in moderate to good yields in the absence of external ligands or bases. The coordination-assisted approach offers the additional benefits that efficient deprotection of the N-substituent and selective aziridine ring-opening are effectively achieved.