MilliporeSigma
  • Synthesis of imidazol-1-yl-acetic acid hydrochloride: a key intermediate for zoledronic acid.
Synthesis of imidazol-1-yl-acetic acid hydrochloride: a key intermediate for zoledronic acid.
Beilstein journal of organic chemistry (2008-12-24)
Santosh Kumar Singh, Narendra Manne, Purna Chandra Ray, Manojit Pal
ABSTRACT
A convenient and practical synthesis of imidazol-1-yl-acetic acid hydrochloride was achieved via N-alkylation of imidazole using tert-butyl chloroacetate followed by a non-aqueous ester cleavage of the resulting imidazol-1-yl-acetic acid tert-butyl ester in the presence of titanium tetrachloride. The synthesized imidazol-1-yl-acetic acid hydrochloride was then utilized to prepare zoledronic acid.
Sigma-Aldrich
tert-Butyl chloroacetate, 97%