MilliporeSigma
  • Effect of processing additive on the nanomorphology of a bulk heterojunction material.
Effect of processing additive on the nanomorphology of a bulk heterojunction material.
Nano letters (2010-09-10)
Ji Sun Moon, Christopher J Takacs, Shinuk Cho, Robert C Coffin, Heejoo Kim, Guillermo C Bazan, Alan J Heeger
ABSTRACT
The bulk heterojunction (BHJ) material Si-PDTBT:PC(70)BM is sensitive to the use of a small amount of 1-chloronaphthalene (CN) as a processing additive; CN as a cosolvent (e.g., 4% in chlorobenzene) causes in a factor of 2 increase in the power conversion efficiency of BHJ solar cells. The morphology of the BHJ material, prepared with and without the CN additive is studied with top-down transmission electron microscopy, cross-sectional transmission electron microscopy, and atomic force microscopy. The improved performance is the result of changes in the nanoscale morphology. Field-effect transistor measurements are consistent with the observed changes in morphology.
Sigma-Aldrich
1-Chloronaphthalene, technical grade
Sigma-Aldrich
1-Chloronaphthalene, technical, ≥85% (GC)