MilliporeSigma
  • Synthesis and antiviral/antitumor evaluation of 2-amino- and 2-carboxamido-3-arylsulfonylthiophenes and related compounds as a new class of diarylsulfones.
Synthesis and antiviral/antitumor evaluation of 2-amino- and 2-carboxamido-3-arylsulfonylthiophenes and related compounds as a new class of diarylsulfones.
Bioorganic & medicinal chemistry (2001-05-30)
C E Stephens, T M Felder, J W Sowell, G Andrei, J Balzarini, R Snoeck, E De Clercq
ABSTRACT
Based on general SARs previously described for anti-HIV-1 diarylsulfone derivatives, a series of 2-amino- and 2-carboxamido-3-arylsulfonylthiophenes has been prepared and evaluated as potential antiviral and antitumor agents. In cell culture, some of the 2-aminothiophenes exhibited moderate and selective activity against HIV-1, with 2-amino-3-(2-nitrophenylsulfonyl)thiophene (7e) being most attractive (EC(50)=3.8 microg/mL, CC(50)=>100 microg/mL). In broad-spectrum antiviral assays, the 3-arylsulfonyl-2-(trifluoroacetamido)thiophenes (8c-g) and 2-acetamido-3-arylsulfonyl-5-nitrothiophenes (9f-g) proved considerably active (IC(50)=0.1-10 microg/mL) against human cytomegalovirus (CMV) and/or varicella zoster virus (VZV). Based on the activity of the trifluoroacetamides, ring-modified furan, N-(substituted)pyrrole, phenyl, and 3,4-thiophene analogues were prepared, and these compounds were also active against CMV and/or VZV, with the notable exception of the 3,4-thiophene derivative. In contrast to other amines, the 2-aminopyrrole precursors (13a-d) also exhibited potent activity against CMV. Unfortunately, most of these compounds displayed significant cytotoxicity against human fibroblasts, the cells supporting CMV and VZV replication, and thus selectivity indices were low. The most notable exception to this was the naphthyl-substituted aminopyrrole 13d, which exhibited both potent (IC(50)=0.3 microg/mL) and selective (CC(50)=>50 microg/mL) activity against CMV. Finally, thiophene aryl amides 8i-k displayed moderate in vitro activity against certain leukemia, breast, and colon cancer cell lines.