MilliporeSigma
  • Isomerization of triphenylmethoxyl and 1,1-diphenylethoxyl radicals. Revised assignment of the electron-spin resonance spectra of purported intermediates formed during the ceric ammonium nitrate mediated photooxidation of aryl carbinols.
Isomerization of triphenylmethoxyl and 1,1-diphenylethoxyl radicals. Revised assignment of the electron-spin resonance spectra of purported intermediates formed during the ceric ammonium nitrate mediated photooxidation of aryl carbinols.
The Journal of organic chemistry (2006-12-16)
K U Ingold, Manuel Smeu, Gino A Dilabio
ABSTRACT
Grossi and Strazzari have reported (J. Org. Chem. 2000, 65, 2748-2754) that the ceric ammonium nitrate modulated photooxidation of triphenylmethanol and 1,1-diphenylethanol yielded ESR spectra of the putative spiro-cyclohexadienyl intermediates in the O-neophyl rearrangements of the corresponding alkoxyl radicals, Ph2(R)CO* (R = Ph, CH3), to the phenoxymethyl radicals, Ph(R)C*OPh. Both ESR spectra are reassigned to the phenoxyl radical, C6H5O*, and the probable mechanism by which phenoxyl is formed in these systems is presented.
Sigma-Aldrich
1,1-Diphenylethanol, 98%