MilliporeSigma
  • Chikungunya outbreak in a rural area of Western Cameroon in 2006: A retrospective serological and entomological survey.
Chikungunya outbreak in a rural area of Western Cameroon in 2006: A retrospective serological and entomological survey.
BMC research notes (2010-05-07)
Maurice Demanou, Christophe Antonio-Nkondjio, Emmanuel Ngapana, Dominique Rousset, Christophe Paupy, Jean-Claude Manuguerra, Hervé Zeller
ABSTRACT
Although arboviral infections including Chikungunya virus (CHIKV) are common in sub-Saharan Africa, data on their circulation and prevalence are poorly documented. In 2006, more than 400 cases of dengue-like fever were reported in Kumbo (Northwest Region of Cameroon). The aim of this study was to identify the aetiology of this fever and to define its extent in the area. We conducted a cross-sectional seroprevalence survey one year after clinical investigations to define the extent of the infection. An entomological survey consisted of the collection and identification of mosquito immature stages in water containers in or around human dwellings. A total of 105 sera were obtained from volunteers and tested for CHIKV, O'Nyong-nyong virus (ONNV) and Dengue virus (DENV) specific IgM and IgG antibodies by enzyme-linked immunosorbent assays (ELISA). CHIKV infection was defined as the presence of IgM antibodies to CHIKV. There was serological evidence for recent Chikungunya infection, as 54 subjects (51.4%) had detectable IgM anti-CHIKV in their sera. Amongst these, 52 showed both anti-CHIKV IgM and IgG, and 2 (1.9%) had IgM anti-CHIKV in the absence of IgG. Isolated anti-CHIKV IgG positives were detected in 41 (39%) cases. No anti-ONNV and anti-DENV IgM antibodies were found amongst the sample tested. Out of 305 larvae collected in the different breeding sites, 87 developed to the adult stage; 56 (64.4%) were Aedes africanus and the remaining Culex spp. These findings suggest that the outbreak of febrile illness reported in three villages of Western Cameroon was due to CHIKV. The issue of a possible persistence of anti-CHIKV IgM antibodies is discussed. Ae. africanus which was found to be relatively abundant among the raffia palm bushes probably plays a role in the transmission of CHIKV along the chain of sylvatic/domestic mosquito species in this rural area. Particular attention should therefore be given to arbovirus infections in the Central African sub-region where these infections are becoming an emerging public health threat.
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody