MilliporeSigma
Microbial D-xylonate production.
Applied microbiology and biotechnology (2012-08-10)
Mervi H Toivari, Yvonne Nygård, Merja Penttilä, Laura Ruohonen, Marilyn G Wiebe
ABSTRACT
D-Xylonic acid is a versatile platform chemical with reported applications as complexing agent or chelator, in dispersal of concrete, and as a precursor for compounds such as co-polyamides, polyesters, hydrogels and 1,2,4-butanetriol. With increasing glucose prices, D-xylonic acid may provide a cheap, non-food derived alternative for gluconic acid, which is widely used (about 80 kton/year) in pharmaceuticals, food products, solvents, adhesives, dyes, paints and polishes. Large-scale production has not been developed, reflecting the current limited market for D-xylonate. D-Xylonic acid occurs naturally, being formed in the first step of oxidative metabolism of D-xylose by some archaea and bacteria via the action of D-xylose or D-glucose dehydrogenases. High extracellular concentrations of D-xylonate have been reported for various bacteria, in particular Gluconobacter oxydans and Pseudomonas putida. High yields of D-xylonate from D-xylose make G. oxydans an attractive choice for biotechnical production. G. oxydans is able to produce D-xylonate directly from plant biomass hydrolysates, but rates and yields are reduced because of sensitivity to hydrolysate inhibitors. Recently, D-xylonate has been produced by the genetically modified bacterium Escherichia coli and yeast Saccharomyces cerevisiae and Kluyveromyces lactis. Expression of NAD(+)-dependent D-xylose dehydrogenase of Caulobacter crescentus in either E. coli or in a robust, hydrolysate-tolerant, industrial Saccharomyces cerevisiae strain has resulted in D-xylonate titres, which are comparable to those seen with G. oxydans, at a volumetric rate approximately 30% of that observed with G. oxydans. With further development, genetically modified microbes may soon provide an alternative for production of D-xylonate at industrial scale.