MilliporeSigma
A concise synthesis of (+/-)-cacalol.
The Journal of organic chemistry (2008-05-30)
Brant L Kedrowski, Robert W Hoppe
ABSTRACT
A simple synthesis of the natural product cacalol has been developed that proceeds in seven steps and 21-25% overall yield. Ortho-lithiation of 4-methylanisole and alkylation with 5-iodo-1-pentene, followed by intramolecular Friedel-Crafts alkylation, gave 5-methoxy-1,8-dimethyltetralin. This compound was then formylated in the 6-position. Baeyer-Villiger oxidation and hydrolysis of the resulting formate gave 6-hydroxy-5-methoxy-1,8-dimethyltetralin. Alkylation of the phenolic hydroxyl group with chloroacetone followed by cyclodehydration gave cacalol methyl ether. Deprotection of this aryl methyl ether yielded cacalol.
Sigma-Aldrich
4-Methylanisole, 99%