MilliporeSigma
Arsenic speciation in rice cereals for infants.
Journal of agricultural and food chemistry (2013-10-02)
Rima Juskelis, Wanxing Li, Jenny Nelson, Jack C Cappozzo
ABSTRACT
The aim of this study was to conduct a survey of arsenic (As) content in rice cereals for infants. The analysis was based on the FDA Elemental Analysis Manual (EAM 4.11). An inductively coupled plasma mass spectrometer (ICP-MS) was used to determine total As. Due to the different toxicities of the chemical forms of arsenic, the ICP-MS coupled to a high-performance liquid chromatograph (HPLC) was used to perform As speciation. The total and speciated arsenic was determined in 31 different infant rice cereals sold in U.S. supermarkets. The mass fraction of total inorganic As (iAs; sum of arsenite As(III) and arsenate As(V)) concentrations ranged between 55.5 ± 1.3 and 158.0 ± 6.0 μg/kg. The average total arsenic and iAs concentrations in infant rice cereal were 174.4 and 101.4 μg/kg, respectively. There was no substantial difference in iAs levels between organic and conventional rice cereals. The mixed-grain rice cereal contained the least total (105 μg/kg) and inorganic arsenic (63 μg/kg). The major detected organoarsenical species was dimethylarsinic acid (DMA). Monomethylarsonic acid (MMA) was not detected, or only trace levels were found. Spiked sample percent recoveries for iAs, DMA, and MMA ranged from a low of 97.3% for iAs to a high of 115.0% for DMA. Results for speciated and total As in the National Institute of Standards and Technology standard reference material rice flour (NIST SRM 1568) were in good agreement with certified values. In the NIST SRM 1568 sample (n = 5) repeatability (%RSD) was 2.8% for iAs, 1.7% for DMA and species sum, and 5.3% for the total arsenic by As total method. The average percent mass balance was 99.9 ± 6.3% for the NIST SRM 1568 sample. This study provides new and much needed information on arsenic levels in rice-based infant cereals.
Product Description
Rice flour (Total As and As species), ERM®, certified reference material