MilliporeSigma
  • Preparation and Characterization of N-Halamine-based Antimicrobial Fillers.
Preparation and Characterization of N-Halamine-based Antimicrobial Fillers.
Industrial & engineering chemistry research (2012-09-04)
Revathi V Padmanabhuni, Jie Luo, Zhengbing Cao, Yuyu Sun
ABSTRACT
The purpose of this study was to demonstrate that the surface of CaCO(3) fillers could be coated with an N-halamine based fatty acid to make the filler surface organophilic and accomplish antibacterial activity simultaneously, rendering the resulting polymer-filler composites antimicrobial. Thus, a new bi-functional compound, 4, 4 -Dimethyl hydantoin-undecanoic acid (DMH-UA), was synthesized by treating the potassium salt of dimethyl hydantoin (DMH) with 11-bromoundecanoic acid (BUA). Upon chlorination treatment with diluted bleach, DMH-UA was transformed into 3-chloro-4, 4-dimethyl hydantoin- undecanoic acid (Cl-DMH-UA). Alternatively, DMH-UA could be coated onto the surface of CaCO(3) to obtain the corresponding calcium salt, 4, 4-dimethyl hydantoin-undecanoic acid-calcium carbonate (DMH-UA-CaCO(3)). In the presence of diluted chlorine bleach, the coated DMH-UA on the surface of CaCO(3) was transformed into Cl-DMH-UA, leading to the formation of Cl-DMH-UA-CaCO(3). The reactions were characterized with FT-IR, NMR, UV, DSC and SEM analyses. Both Cl-DMH-UA and Cl-DMH-UA-CaCO(3) were used as antimicrobial additives for cellulose acetate (CA). The antimicrobial efficacy of the resulting samples was evaluated against both Escherichia coli (Gram-negative bacteria) and Staphylococcus aureus (Gram-positive bacteria). It was found that with the same additive content, CA samples with Cl-DMH-UA-CaCO(3) and Cl-DMH-UA had very similar antimicrobial and biofilm-controlling activity, but the former released less active chlorine into the surrounding environment than the latter.
Sigma-Aldrich
11-Bromoundecanoic acid, technical grade, 95%