MilliporeSigma
  • High-yield radiosynthesis and preliminary in vivo evaluation of p-[18F]MPPF, a fluoro analog of WAY-100635.
High-yield radiosynthesis and preliminary in vivo evaluation of p-[18F]MPPF, a fluoro analog of WAY-100635.
Nuclear medicine and biology (1998-06-25)
D Le Bars, C Lemaire, N Ginovart, A Plenevaux, J Aerts, C Brihaye, W Hassoun, V Leviel, P Mekhsian, D Weissmann, J F Pujol, A Luxen, D Comar
ABSTRACT
No-carrier-added 4-[18F]fluoro-N-[2-[1-(2-methoxyphenyl)-1 piperazinyl]ethyl-N-2-pyridinyl-benzamide (p-[18F]MPPF) was synthesized by nucleophilic substitution of the corresponding nitro compound in the presence of Kryptofix 222 and K2CO3 by microwave heating (3 min, 500 W) using a remotely controlled radiosynthesis. Baseline separation of p-[18F]MPPF from the nitro derivative was performed on a semipreparative HPLC C18 column. After Sep-Pak formulation, the radiopharmaceutical was obtained with a radiochemical yield of 25% (EOS) in about 70 min. Specific radioactivity averaged between 1-5 Ci/micromol EOS. Labelling of the ortho and meta derivatives was also attempted. Brain uptake of p-[18F]MPPF was studied with PET on fluothane-anesthetized cats. Following intravenous injection of p-[18F]MPPF, high accumulation of radioactivity was observed in the hippocampus and cerebral cortex. Low levels of radioactivity were observed in cerebellum. At 30 min, the mean hippocampus/cerebellum and cortex/cerebellum ratios were 5 and 3.8, respectively. The accumulation of the tracer was blocked by prior administration of reference WAY-100635, demonstrating the specificity of the ligand.