MilliporeSigma
  • High capacity carbon-coated Si70Sn30 nanoalloys for lithium battery anode material.
High capacity carbon-coated Si70Sn30 nanoalloys for lithium battery anode material.
Chemical communications (Cambridge, England) (2008-02-23)
YooJeong Kwon, Jaephil Cho
ABSTRACT
Carbon-coated Si70Sn30 nanoalloys with a particle size < 10 nm were prepared from butyl-capped analogues via firing at 900 degrees C under a vacuum showed a reversible capacity of 2032 mAh g(-1) and excellent capacity retention.
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Naphthalene, 99%