MilliporeSigma
  • Panoramic imaging of the temporomandibular joint: an experimental study using cadaveric skulls.
Panoramic imaging of the temporomandibular joint: an experimental study using cadaveric skulls.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2006-01-18)
Steven D Fallon, Gabriel W Fritz, Daniel M Laskin
ABSTRACT
The purpose of this study was to determine if condylar morphology can be accurately depicted on the panoramic radiograph and if variations in condylar anatomy affect the radiographic appearance of the condyle. The shape and angulation of the condyle were determined on 8 cadaveric human skulls. Lead foil markers were then placed on the articulating surface and lateral and medial pole of each condyle. A panoramic radiograph was made of each skull and the position of the markers was assessed. The true shape of the condyles was not recognizable on any of the panoramic images. The exact location of the articulating surface and medial and lateral poles could not be predicted on the panoramic images without markers. The location of these structures varied depending on the amount of condylar angulation. Because of the radiographic variations produced by differences in condylar angulation, it is not possible to accurately determine condylar morphology on the panoramic radiograph. Therefore, its value in providing the detailed information needed to guide diagnosis and treatment of the temporomandibular joint patient is limited.
Product Description
Lead, foil, 4mm disks, thickness 0.009mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.004mm, 99.99+%
Lead, foil, 2m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 4mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.006mm, 99.99+%
Lead, foil, 1m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 1m coil, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 0.5m coil, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 6mm disks, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 0.5m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.03mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 0.5m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 15mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.03mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 8mm disks, thickness 0.006mm, 99.99+%
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 3.0mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 100x100mm, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 20m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%