MilliporeSigma
  • Recognition of sequence-information in synthetic copolymer chains by a conformationally-constrained tweezer molecule.
Recognition of sequence-information in synthetic copolymer chains by a conformationally-constrained tweezer molecule.
Faraday discussions (2009-01-01)
Howard M Colquhoun, Zhixue Zhu, Christine J Cardin, Michael G B Drew, Yu Gan
ABSTRACT
A novel type of tweezer molecule containing electron-rich 2-pyrenyloxy arms has been designed to exploit intramolecular hydrogen bonding in stabilising a preferred conformation for supramolecular complexation to complementary sequences in aromatic copolyimides. This tweezer-conformation is demonstrated by single-crystal X-ray analyses of the tweezer molecule itself and of its complex with an aromatic diimide model-compound. In terms of its ability to bind selectively to polyimide chains, the new tweezer molecule shows very high sensitivity to sequence effects. Thus, even low concentrations of tweezer relative to diimide units (< 2.5 mol %) are sufficient to produce dramatic, sequence-related splittings of the pyromellitimide proton NMR resonances. These induced resonance-shifts arise from ring-current shielding of pyromellitimide protons by the pyrenyloxy arms of the tweezer-molecule, and the magnitude of such shielding is a function of the tweezer-binding constant for any particular monomer sequence. Recognition of both short-range and long-range sequences is observed, the latter arising from cumulative ring-current shielding of diimide protons by tweezer molecules binding at multiple adjacent sites on the copolymer chain.
Sigma-Aldrich
2,6-Pyridinedicarbonyl dichloride, 97%
Sigma-Aldrich
4-Amino-3-methylphenol, 97%