MilliporeSigma
  • Synthesis and carbonic anhydrase inhibitory properties of sulfamides structurally related to dopamine.
Synthesis and carbonic anhydrase inhibitory properties of sulfamides structurally related to dopamine.
Bioorganic & medicinal chemistry (2013-04-30)
Kadir Aksu, Meryem Nar, Muhammet Tanc, Daniela Vullo, Ilhami Gülçin, Süleyman Göksu, Ferhan Tümer, Claudiu T Supuran
ABSTRACT
A series of novel sulfamides incorporating the dopamine scaffold were synthesized. Reaction of amines and tert-butyl-alcohol/benzyl alcohol in the presence of chlorosulfonyl isocyanate (CSI) afforded sulfamoyl carbamates, which were converted to the title compounds by treatment with trifluoroacetic acid or by palladium-catalyzed hydrogenolysis. Inhibition of six α-carbonic anhydrases (CAs, EC 4.2.1.1), that is, CA I, CA II, CA VA, CA IX, CA XII and CA XIV, and two β-CAs from Candida glabrata (CgCA) and Mycobacterium tuberculosis (Rv3588) with these sulfamides was investigated. All CA isozymes were inhibited in the low micromolar to nanomolar range by the dopamine sulfamide analogues. K(i)s were in the range of 0.061-1.822 μM for CA I, 1.47-2.94 nM for CA II, 2.25-3.34 μM for CA VA, 0.041-0.37 μM for CA IX, 0.021-1.52 μM for CA XII, 0.007-0.219 μM for CA XIV, 0.35-5.31 μM for CgCA and 0.465-4.29 μM for Rv3588. The synthesized sulfamides may lead to inhibitors targeting medicinally relevant CA isoforms with potential applications as antiepileptic, antiobesity antitumor agents or anti-infective.
Sigma-Aldrich
Chlorosulfonyl isocyanate, 98%