MilliporeSigma
  • Inhibition of Hageman factor, plasma thromboplastin antecedent, thrombin and other clotting factors by phenylglyoxal hydrate (38500).
Inhibition of Hageman factor, plasma thromboplastin antecedent, thrombin and other clotting factors by phenylglyoxal hydrate (38500).
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.) (1975-01-01)
O D Radnoff, H Saito
PMID236567
ABSTRACT
Exposure of purified Hageman factor (HF, Factor XII) to phenylglyoxal hydrate (PHG), an agent reacting with arginine residues in protein, inhibited its coagulant properties upon subsequent exposure of negatively charged agents. Once HF had been exposed to kaolin or ellagic acid, however, subsequent addition of PHG was much less inhibitory. PHG had no effect upon the ability of HF to bind to negatively charged surfaces. PGH also inhibited preparations of activated PTA (Factor XI) and thrombin, and, when incubated with plasma, reduced the titer of coagulable fibrinogen, PTA Christmas factor (Factor IX), antihemophilic factor (Factor VIII), Factor VII, Stuart factor (Factor X), proaccelerin (Factor V) and prothrombin (Factor II), and to a lesser degres, HF.
Sigma-Aldrich
Phenylglyoxal hydrate, 97%