MilliporeSigma
  • Design and synthesis of new biprivileged molecular scaffolds: indolo-fused benzodiazepinyl/quinoxalinyl benzimidazoles.
Design and synthesis of new biprivileged molecular scaffolds: indolo-fused benzodiazepinyl/quinoxalinyl benzimidazoles.
Chemistry, an Asian journal (2012-04-19)
Indrajeet J Barve, Chan-Yu Chen, Deepak B Salunke, Wen-Sheng Chung, Chung-Ming Sun
ABSTRACT
The present article describes the design and synthesis of new biprivileged molecular scaffolds with diverse structural features. Commercially available, simple heterocyclic building blocks such as 4-fluoro-3-nitrobenzoic acid, 2-chloro-3-nitrobenzoic acid, and indoline were utilized for the synthesis of the novel heterocycles. Pictet-Spengler-type condensation was used as a key step to construct tetracyclic indolo-benzodiazepines and indolo-quinoxalines linked with substituted benzimidazoles. Analysis of single crystals of representative compounds showed that these molecular skeletons have the potential to present various substituents with distinct three-dimensional orientations.