MilliporeSigma
  • Catalytic hydrogenation activity and electronic structure determination of bis(arylimidazol-2-ylidene)pyridine cobalt alkyl and hydride complexes.
Catalytic hydrogenation activity and electronic structure determination of bis(arylimidazol-2-ylidene)pyridine cobalt alkyl and hydride complexes.
Journal of the American Chemical Society (2013-08-24)
Renyuan Pony Yu, Jonathan M Darmon, Carsten Milsmann, Grant W Margulieux, S Chantal E Stieber, Serena DeBeer, Paul J Chirik
ABSTRACT
The bis(arylimidazol-2-ylidene)pyridine cobalt methyl complex, ((iPr)CNC)CoCH3, was evaluated for the catalytic hydrogenation of alkenes. At 22 °C and 4 atm of H2 pressure, ((iPr)CNC)CoCH3 is an effective precatalyst for the hydrogenation of sterically hindered, unactivated alkenes such as trans-methylstilbene, 1-methyl-1-cyclohexene, and 2,3-dimethyl-2-butene, representing one of the most active cobalt hydrogenation catalysts reported to date. Preparation of the cobalt hydride complex, ((iPr)CNC)CoH, was accomplished by hydrogenation of ((iPr)CNC)CoCH3. Over the course of 3 h at 22 °C, migration of the metal hydride to the 4-position of the pyridine ring yielded (4-H2-(iPr)CNC)CoN2. Similar alkyl migration was observed upon treatment of ((iPr)CNC)CoH with 1,1-diphenylethylene. This reactivity raised the question as to whether this class of chelate is redox-active, engaging in radical chemistry with the cobalt center. A combination of structural, spectroscopic, and computational studies was conducted and provided definitive evidence for bis(arylimidazol-2-ylidene)pyridine radicals in reduced cobalt chemistry. Spin density calculations established that the radicals were localized on the pyridine ring, accounting for the observed reactivity, and suggest that a wide family of pyridine-based pincers may also be redox-active.
Sigma-Aldrich
1-Methyl-1-cyclohexene, 97%