MilliporeSigma
  • A method for the quantitative recovery of protein in dilute solution in the presence of detergents and lipids.
A method for the quantitative recovery of protein in dilute solution in the presence of detergents and lipids.
Analytical biochemistry (1984-04-01)
D Wessel, U I Flügge
ABSTRACT
A rapid method based on a defined methanol-chloroform-water mixture for the quantitative precipitation of soluble as well as hydrophobic proteins from dilute solutions (e.g., column chromatography effluents) has been developed. The effectiveness of this method is not affected by the presence of detergents, lipids, salt, buffers, and beta-mercaptoethanol.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets