MilliporeSigma
  • Synthesis and biological evaluation of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid as a potential inhibitor of GAR Tfase and the de novo purine biosynthetic pathway.
Synthesis and biological evaluation of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid as a potential inhibitor of GAR Tfase and the de novo purine biosynthetic pathway.
Bioorganic & medicinal chemistry (2005-04-26)
Heng Cheng, Inkyu Hwang, Youhoon Chong, Ali Tavassoli, Michael E Webb, Yan Zhang, Ian A Wilson, Stephen J Benkovic, Dale L Boger
ABSTRACT
The synthesis and evaluation of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid (2) as an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase) are reported. The inhibitor 2 was prepared in a convergent synthesis involving C-alkylation of methyl 4-(4,4,4-trifluoro-3-dimethylhydrazonobutyl)benzoate with 1-chloro-3-iodopropane followed by construction of the pyrimidinone ring. Compound 2 was found to be an effective inhibitor of recombinant human GAR Tfase (K(i) = 0.50 microM), whereas it was inactive (K(i) > 100 microM) against E. coli GAR Tfase as well as recombinant human AICAR Tfase. Compound 2 exhibited modest, purine-sensitive growth inhibitory activity against the CCRF-CEM cell line (IC50 = 6.0 microM).
Sigma-Aldrich
1-Chloro-3-iodopropane, 99%
CCRF-CEM, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85112105, human blood, Lymphoblast