MilliporeSigma
  • Synthesis and cytotoxic activity of benzophenanthrolinone analogues of acronycine.
Synthesis and cytotoxic activity of benzophenanthrolinone analogues of acronycine.
Chemical & pharmaceutical bulletin (2001-09-18)
J B Bongui, A Elomri, E Seguin, F Tillequin, B Pfeiffer, P Renard, A Pierré, G Atassi
ABSTRACT
Condensation of either 2-bromobenzoic acid (4) or 2-chloro-3-nitrobenzoic acid (5) with suitable aminoquinolines 6-8 afforded phenylquinolylamines 9-13. Acid mediated cyclization gave the corresponding 12H-benzo[b][1,7]phenanthrolin-7-ones 14 and 15, and 12H-benzo[b][1,10]phenanthrolin-7-ones 16-18. Compounds 14, 16, and 17 were subsequently N-methylated to 6-demethoxyacronycine and acronycine analogues 19-21, whereas reduction of the aromatic nitro group of 18 gave the amino derivative 22. Unsubstituted 12H-benzo[b][1,10]phenanthrolin-7-ones 16, 17, 20, and 21 were devoid of significant cytotoxic activity, whereas 18 and 22, bearing a nitrogen substituent at position 11, were significantly active. Unsubstituted 12H-benzo[b][1,7]phenanthrolin-7-ones 14 and 19, which include a pyridine nitrogen in the same 4-position as the pyran oxygen of acronycine exhibited cytotoxic activities within the same range of magnitude as acronycine itself.
Sigma-Aldrich
2-Bromobenzoic acid, 97%
Pricing and availability is not currently available.