MilliporeSigma
  • Comparison of subcritical water and organic solvents for extracting kava lactones from kava root.
Comparison of subcritical water and organic solvents for extracting kava lactones from kava root.
Journal of chromatography. A (2001-08-21)
A Kubátová, D J Miller, S B Hawthorne
ABSTRACT
Subcritical water extraction of lactones from a kava (Piper metlhysticum) root was compared to a Soxhlet extraction with water, to boiling in water, and to a sonication in acetone. For ground kava (250-500 microm), 2 h of subcritical water extraction were required for a complete extraction at 100 degrees C, while at 175 degrees C, 20 min were sufficient. For a complete extraction of the unground (shredded) kava, the time of extraction was extended to 40 min at 175 degrees C. Boiling for 2 h and extraction with Soxhlet apparatus for 6 h, both of which employed water at atmospheric pressure, produced yields 40-60% lower than those obtained with subcritical water. With unground kava, 40 min of subcritical water extraction yielded essentially the same recoveries of lactones as 18 h of sonication with acetone, methylene chloride, or methanol.
Sigma-Aldrich
Heptadecane, 99%