MilliporeSigma
  • Krüppel-like factor KLF9 regulates PPARγ transactivation at the middle stage of adipogenesis.
Krüppel-like factor KLF9 regulates PPARγ transactivation at the middle stage of adipogenesis.
Cell death and differentiation (2010-08-21)
H Pei, Y Yao, Y Yang, K Liao, J-R Wu
ABSTRACT
Krüppel-like factors (KLFs) as a family of zinc-finger transcription factors involve in the regulation of many physiological processes. In these studies, KLF9 was characterized for its role in adipogenesis. The expression of KLF9 was markedly upregulated during the middle stage of 3T3-L1 adipocyte differentiation, and inhibition of KLF9 by RNAi impaired adipogenesis. Using promoter deletion and mutation analysis, we identified two KLF9-binding sites within the 0.6-kb region of the PPARγ2 proximal promoter, indicating that KLF9 interacts with the PPARγ2 promoter. Furthermore, we found that KLF9 could synergistically activate PPARγ2 promoter by directly interacting with C/EBPα. In addition, overexpression of PPARγ2 rescued the impairment of adipocyte differentiation induced by KLF9 knockdown, which supports that PPARγ2 is a downstream target of KLF9. Collectively, our results indicate KLF9 as a key pro-adipogenic transcription factor through regulation of PPARγ2 expression with C/EBPα at the middle stage of adipogenesis.
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture